missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ IsoClean™ Zipper Front Cleanroom Frocks
GREENER_CHOICE

Catalog No. 17987101
Change view
missing translation for 'orderingAttributeHoverText'
Size:
2X-Large
3X-Large
4X-Large
Large
Medium
Small
X-Large
Length (English):
41.5 in.
43.75 in.
45.75 in.
47.75 in.
53.75 in.
55 in.
57 in.
Size (English) Chest:
28 to 32 in.
32 to 36 in.
36 to 38 in.
38 to 40 in.
40 to 43 in.
43 to 46 in.
46 to 52 in.
Catalog No. Size Length (English) Size (English) Chest
17987101 4X-Large 57 in. 46 to 52 in.
17988081 Medium 43.75 in. 32 to 36 in.
17988084 2X-Large 53.75 in. 40 to 43 in.
17988083 X-Large 47.75 in. 38 to 40 in.
17988082 Large 45.75 in. 36 to 38 in.
17988080 Small 41.5 in. 28 to 32 in.
17987100 3X-Large 55 in. 43 to 46 in.
7 options
Should not be worn around open flame, heat source, sparks or ignition sources.

Catalog No. 17987101

Supplier: DuPont™ IC264SWH4X00300B

Only null left
Should not be worn around open flame, heat source, sparks or ignition sources.

Frock provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Bulk packaged in double transparent poly liners.

Frock provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. ulk packaged in double transparent poly liners.

  • Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam
  • Elastic openings for tighter fit at wrist
  • Bound neck for lower particle shedding
  • Front zipper closure for easy donning and doffing
  • Raglan sleeve design for greater range of motion and comfort
  • A-line silhouette

Specifications

Product Type Controlled Environments Frock
Size 4X-Large
Material Tyvek
Color White
Sterility Non-sterile
Closure Type Zipper
Cuff Style Elastic
Seam Type Serged
Collar Type Bound
Size (English) Chest 46 to 52 in.
Size (Metric) Chest 116.84 to 132.08 cm
Sleeve Style Raglan Sleeve
Packaging Type Bulk Packaged
Length (English) 57 in.
Length (Metric) 144.78 cm
Length (English) Sleeve 46 in.
Length (Metric) Sleeve 116.84 cm
Cleanroom Class Class 1000 (ISO 6)
Show More Show Less