Learn More
DuPont™ Tyvek™ IsoClean™ Clean-Processed and Sterile
Lightweight, breathable, and clean processed coveralls for use in ISO 4-5 Class 10-100 critical environments.
Supplier:  DuPont™ IC254BWH3X0025CS
 View more versions of this product
Catalog No. 17-985-405
Only null left
Explore available promotions
Description
Description
For use in controlled environments with high standards for particle and microbiological contamination control (pharmaceutical, biotech, medical device and electronic settings).
  • Material and bound seams offer an excellent barrier while maintaining comfort and durability
  • Gamma sterilized and clean processed
  • Spun polyethylene creates barrier to particles, micro-organisms and non-hazardous liquids
  • Lowest level particle shedding
  • Bound neck and seams
  • Elastic wrists and ankles
  • Dolman sleeves
  • Zipper closure
Should not be worn around open flame, heat source, sparks or ignition sources
Specifications
Specifications
Cleanroom Coveralls
Unisex
White
Elastic
Collared
Bound
Anti-static, Abrasion
Yes
35 in.
88.9 cm
104.7 cm
6.16 to 6.5 ft.
Class 10-100 (ISO 4-5)
3X-Large
Tyvek Flashspun Polyethylene
Single-use
Elastic
Zipper
Sterile
Bound
46 to 52 in.
116.8 to 132 cm
41.25 in.
30.5 in.
187.9 to 198.1 cm
Product Suggestions
Product Suggestions