missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ IsoClean™ Clean-Processed and Sterile
GREENER_CHOICE

Catalog No. 17985404
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17985404 2X-Large
17985405 3X-Large
17985401 Medium
17985403 X-Large
17985406 4X-Large
17985400 Small
17985402 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 17985404 Supplier DuPont™ Supplier No. IC254BWH2X0025CS

Lightweight, breathable, and clean processed coveralls for use in ISO 4-5 Class 10-100 critical environments.

For use in controlled environments with high standards for particle and microbiological contamination control (pharmaceutical, biotech, medical device and electronic settings).

  • Material and bound seams offer an excellent barrier while maintaining comfort and durability
  • Gamma sterilized and clean processed
  • Spun polyethylene creates barrier to particles, micro-organisms and non-hazardous liquids
  • Lowest level particle shedding
  • Bound neck and seams
  • Elastic wrists and ankles
  • Dolman sleeves
  • Zipper closure

Should not be worn around open flame, heat source, sparks or ignition sources

Specifications

Product Type Cleanroom Coveralls
Size 2X-Large
Gender Unisex
Material Tyvek Flashspun Polyethylene
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Collared
Closure Type Zipper
Seam Type Bound
Sterility Sterile
Resistance Anti-static, Abrasion
Latex Free Yes
Size (English) Chest 43 to 46 in.
Size (English) Inseam 34 in.
Size (Metric) Chest 109.2 to 116.8 cm
Size (Metric) Inseam 86.3 cm
Length (English) Sleeve 40.25 in.
Length (Metric) Sleeve 102.2 cm
Width (English) Chest 28.5 in.
Height (English) 6 to 6.4 ft.
Height (Metric) 185.4 to 195.5 cm
Cleanroom Class Class 10-100 (ISO 4-5)
Show More Show Less