missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™ IsoClean™ Clean-Processed and Sterile
Lightweight, breathable, and clean processed coveralls for use in ISO 4-5 Class 10-100 critical environments.
Supplier: DuPont™ IC254BWHLG0025CS
Description
For use in controlled environments with high standards for particle and microbiological contamination control (pharmaceutical, biotech, medical device and electronic settings).
- Material and bound seams offer an excellent barrier while maintaining comfort and durability
- Gamma sterilized and clean processed
- Spun polyethylene creates barrier to particles, micro-organisms and non-hazardous liquids
- Lowest level particle shedding
- Bound neck and seams
- Elastic wrists and ankles
- Dolman sleeves
- Zipper closure
Should not be worn around open flame, heat source, sparks or ignition sources
Specifications
| Cleanroom Coveralls | |
| Unisex | |
| White | |
| Elastic | |
| Collared | |
| Bound | |
| Anti-static, Abrasion | |
| 38 to 40 in. | |
| 81.9 cm | |
| 94.4 cm | |
| 5.6 to 6 ft. |
| Large | |
| Tyvek Flashspun Polyethylene | |
| Single-use | |
| Elastic | |
| Zipper | |
| Sterile | |
| Yes | |
| 96.5 to 101.6 cm | |
| 37.25 in. | |
| 24.5 in. | |
| Class 10-100 (ISO 4-5) |