missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™ IsoClean™ Coverall (all)

Description
- For use in controlled environments with high standards for particle and microbiological contamination control:
- Pharmaceutical
- Biotech
- Medical Device
- Electronics
- Light and durable
- Spun polyethylene creates barrier to particles, micro-organisms, and non-hazardous liquids
- Lowest level particle shedding
- Bound seams and neck
- Covered elastic wrists and ankles with dolman sleeves
- Zipper closure
- Should not be worn around open flame, heat source, sparks or ignition sources
Specifications
Specifications
| Product Type | Cleanroom Coveralls |
| Size | 2X-Large |
| Gender | Unisex |
| Material | Tyvek |
| Color | White |
| Disposable | Single-use |
| Waist Style | Non-elastic |
| Ankle Style | Covered Elastic |
| Cuff Style | Covered Elastic |
| Neck Style | Collared |
| Show More |