Learn More
DuPont™ Tyvek™ IsoClean™ Coverall (all)
Tyvek coveralls provide an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash. Bound seams reduce the potential for particle penetration.
Supplier:  DuPont™ IC253BWHMD00250B
 View more versions of this product
Catalog No. 17-987-992
Only null left
Explore available promotions
Alert:
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Description
Description
  • For use in controlled environments with high standards for particle and microbiological contamination control:
    • Pharmaceutical
    • Biotech
    • Medical Device
    • Electronics
  • Light and durable
  • Spun polyethylene creates barrier to particles, micro-organisms, and non-hazardous liquids
  • Lowest level particle shedding
  • Bound seams and neck
  • Covered elastic wrists and ankles with dolman sleeves
  • Zipper closure
  • Should not be worn around open flame, heat source, sparks or ignition sources
Specifications
Specifications
Cleanroom Coveralls
Unisex
White
Covered Elastic
Collared
Bound
Bound
30.5 in.
77.4 cm
5.4 mil
35.5 in.
5.5 to 5.10 ft.
Bulk
Medium
Tyvek
Single-use
Elastic
Zipper
Non-sterile
36 to 38 in.
91.44 to 96.52 cm
Dolman
Barrier to Dry Particles, Microorganisms, and Non-Hazardous Liquids in Controlled Environments
90.17 cm
167.64 to 155.44 cm
Class 1000 (ISO 6)
Product Suggestions
Product Suggestions