missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ IsoClean™ Coverall (all)
GREENER_CHOICE

Catalog No. 17987991
Change view
Click to view available options
Size:
Small
Medium
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Height (English):
5 to 5.7 ft.
5.11 to 6.3 ft.
5.5 to 5.10 ft.
6.1 to 6.5 ft.
6.2 to 6.6 ft.
6.3 to 7 ft.
6.4 to 7.2 ft.
Height (Metric):
152.4 to 173.73 cm
155.75 to 192.04 cm
167.64 to 155.44 cm
185.92 to 198.12 cm
188.97 to 201.16 cm
192.02 to 213.3 cm
195.07 to 219.45 cm
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Height (English) Height (Metric)
17987991 Small 5 to 5.7 ft. 152.4 to 173.73 cm
17987992 Medium 5.5 to 5.10 ft. 167.64 to 155.44 cm
17987994 X-Large 5.11 to 6.3 ft. 155.75 to 192.04 cm
17987995 2X-Large 6.1 to 6.5 ft. 185.92 to 198.12 cm
17987996 3X-Large 6.2 to 6.6 ft. 188.97 to 201.16 cm
17987997 4X-Large 6.3 to 7 ft. 192.02 to 213.3 cm
17987998 5X-Large 6.4 to 7.2 ft. 195.07 to 219.45 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Catalog No. 17987991 Supplier DuPont™ Supplier No. IC253BWHSM00250B
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.

Tyvek coveralls provide an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash. Bound seams reduce the potential for particle penetration.

  • For use in controlled environments with high standards for particle and microbiological contamination control:
    • Pharmaceutical
    • Biotech
    • Medical Device
    • Electronics
  • Light and durable
  • Spun polyethylene creates barrier to particles, micro-organisms, and non-hazardous liquids
  • Lowest level particle shedding
  • Bound seams and neck
  • Covered elastic wrists and ankles with dolman sleeves
  • Zipper closure
  • Should not be worn around open flame, heat source, sparks or ignition sources

Specifications

Product Type Cleanroom Coveralls
Size Small
Gender Unisex
Material Tyvek
Color White
Disposable Single-use
Ankle Style Covered Elastic
Cuff Style Covered Elastic
Neck Style Collared
Closure Type Zipper
Seam Type Bound
Sterility Non-sterile
Collar Type Bound
Size (English) Chest 32 to 36 in.
Size (English) Inseam 29.25 in.
Size (Metric) Chest 81.28 to 91.44 cm
Size (Metric) Inseam 74.2 cm
Sleeve Style Dolman
Thickness 5.4 mil
For Use With (Application) Barrier to Dry Particles, Microorganisms, and Non-Hazardous Liquids in Controlled Environments
Length (English) Sleeve 34.25 in.
Length (Metric) Sleeve 86.9 cm
Height (English) 5 to 5.7 ft.
Height (Metric) 152.4 to 173.73 cm
Packaging Bulk
Cleanroom Class Class 1000 (ISO 6)
Show More Show Less