missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ IsoClean™ Coverall (all)
GREENER_CHOICE

Catalog No. 19067015
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19067015 5X-Large
17988135 Small
17988037 Medium
17988038 Large
17988039 X-Large
17988040 2X-Large
17988041 3X-Large
17988042 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Catalog No. 19067015 Supplier DuPont™ Supplier No. IC253BWH5X00250S

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.

Sterile coveralls used in controlled environments to protect against particulates and other contaminants.

  • Garments are gamma sterilized to SAL of 10-6
  • Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration
  • Coverall has elastic openings for tighter fit at wrist and ankle
  • Bound neck for lower particle shedding
  • Front zipper closure for easy donning and doffing
  • Dolman sleeve design for greater range of motion and comfort
  • Full traceability on all sterilized apparel with certificates of sterility available

Specifications

Product Type Cleanroom Coveralls
Size 5X-Large
Gender Unisex
Material Tyvek
Color White
Disposable Single-use
Waist Style Non-elastic
Ankle Style Covered Elastic
Cuff Style Elastic
Neck Style Collared
Hood Type Standard Collar
Closure Type Zipper
Seam Type Bound
Sterility Sterile
Resistance Particle, Non-toxic Particulates
Model IC253BWH
Thickness 5 mil
For Use With (Application) Barrier to Dry Particles, Microorganisms and Non-Hazardous Liquids
Width (English) Chest 34.25 in.
Height (English) 6 ft. 7 in. to 6 ft. 10 in.
Cleanroom Class Class 100-100,000 (ISO 5-8)
Show More Show Less