Learn More
DuPont™ Tyvek™ IsoClean™ Coverall (all)
Sterile coveralls used in controlled environments to protect against particulates and other contaminants.
Supplier:  DuPont™ IC253BWHLG00250S
 View more versions of this product
Catalog No. 17-988-038
Only null left
Explore available promotions
Alert:
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Description
Description
  • Garments are gamma sterilized to SAL of 10-6
  • Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration
  • Coverall has elastic openings for tighter fit at wrist and ankle
  • Bound neck for lower particle shedding
  • Front zipper closure for easy donning and doffing
  • Dolman sleeve design for greater range of motion and comfort
  • Full traceability on all sterilized apparel with certificates of sterility available
Specifications
Specifications
Cleanroom Coveralls
Unisex
White
Covered Elastic
Zipper
Sterile
IC253BWH
32.25 in.
81.9 cm
5 mil
36.75 in.
5.8 to 6 ft.
Large
Tyvek
Covered Elastic
Collared
Bound
Bound
38 to 40 in.
96.52 to 101.6 cm
Dolman
Barrier to Dry Particles, Microorganisms, and Non-Hazardous Liquids
93.35 cm
176.78 to 182.88 cm
Product Suggestions
Product Suggestions