Learn More
DuPont™ Tyvek™ IsoClean™ Lab Coat
Protect against particulates and other contaminants in controlled environments.
Supplier:  DuPont™ IC224SWHXL00300B
 View more versions of this product
Catalog No. 17988025
Only null left
Explore available promotions
Alert:
Should not be worn around open flame, heat source, sparks or ignition sources.
Description
Description
  • Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam.
  • Laydown collar
  • Labcoat offers front snap closures for easy donning and doffing
  • Left chest pocket and two lower pockets
  • Raglan sleeve design for greater range of motion and comfort.
  • Bulk packaged in double transparent poly liners.
  • Manufactured under specifications that do not contain natural rubber latex
Specifications
Specifications
Controlled Environments Lab Coat
Tyvek
Non-sterile
45 in.
Class 1000 (ISO 6)
Unisex
Single-use
Raglan
114.3 cm
Product Suggestions
Product Suggestions