Learn More
DuPont™ Tyvek™ IsoClean™ Lab Coat
GREENER_CHOICE
Catalog No. 17988024
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17988024 Large
17988026 2X-Large
17988001 Medium
17988000 -
17988025 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Should not be worn around open flame, heat source, sparks or ignition sources.
Catalog No. 17988024 Supplier DuPont™ Supplier No. IC224SWHLG00300B
Only null left
Should not be worn around open flame, heat source, sparks or ignition sources.
Protect against particulates and other contaminants in controlled environments.
  • Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam.
  • Laydown collar
  • Labcoat offers front snap closures for easy donning and doffing
  • Left chest pocket and two lower pockets
  • Raglan sleeve design for greater range of motion and comfort.
  • Bulk packaged in double transparent poly liners.
  • Manufactured under specifications that do not contain natural rubber latex
Specifications
Product Type Controlled Environments Lab Coat
Size Large
Gender Unisex
Material Tyvek
Color White
Closure Type Snaps
Cuff Style Open
Disposable Single-use
Sterility Non-sterile
Collar Type Lay Down
Seam Type Serged
Length (English) 43.5 in.
Length (Metric) 110.4 cm
Cleanroom Class Class 1000 (ISO 6)
Show More Show Less