missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ IsoClean™ Series 180 Coveralls, Attached Hood
GREENER_CHOICE

Catalog No. 17978124
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Size (Metric) Chest:
101.6 to 109.2 cm
109.2 to 116.8 cm
116.8 to 132 cm
91.4 to 96.5 cm
96.5 to 101.6 cm
Length (Metric) Sleeve:
84.4 cm
88.9 cm
92.7 cm
97.1 cm
97.7 cm
Size (Metric) Inseam:
74.9 cm
77.47 cm
80 cm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Size (Metric) Chest Length (Metric) Sleeve Size (Metric) Inseam
17978124 3X-Large 116.8 to 132 cm 97.7 cm 80 cm
17978123 2X-Large 109.2 to 116.8 cm 97.1 cm 77.47 cm
17978120 Medium 91.4 to 96.5 cm 84.4 cm -
17978121 Large 96.5 to 101.6 cm 88.9 cm -
17978122 X-Large 101.6 to 109.2 cm 92.7 cm 74.9 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Should not be worn around open flame, heat source, sparks or ignition sources.
Catalog No. 17978124 Supplier DuPont™ Supplier No. IC180SWH3X002500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Should not be worn around open flame, heat source, sparks or ignition sources.

Light and durable coveralls provide a barrier to particles, microorganisms, and non-hazardous light liquid splash.

  • IsoClean protects against micro-organisms and particulates.
  • Spinning process creates barrier to particles, micro-organisms, and nonhazardous liquids
  • Lowest level particle shedding
  • Recyclable
  • Serged seams
  • Standard hood
  • Elastic wrists and ankles
  • Zipper closure
  • Use in controlled environments with high standards for particle and microbiological contamination control
  • Pharmaceutical, biotech, medical device, electronic settings

Specifications

Product Type Cleanroom Coveralls
Size 3X-Large
Gender Unisex
Material Tyvek
Color White
Disposable Single-use
Waist Style Non-elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Attached, Elastic Face
Closure Type Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Tears
Latex Free Yes
Size (English) Chest 46 to 52 in.
Size (English) Inseam 31.5 in.
Size (Metric) Chest 116.8 to 132 cm
Size (Metric) Inseam 80 cm
Length (English) Sleeve 38.5 in.
Length (Metric) Sleeve 97.7 cm
Width (English) Chest 30.25 in.
Packaging Bulk
Cleanroom Class Class 1000-100,000 (ISO 6-8)
Show More Show Less