missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™ IsoClean™ Series 180 Coveralls, Attached Hood
Light and durable coveralls provide a barrier to particles, microorganisms, and non-hazardous light liquid splash.
Supplier: DuPont™ IC180SWHMD002500
Alert:
Should not be worn around open flame, heat source, sparks or ignition sources.
Description
IsoClean protects against micro-organisms and particulates.
- Spinning process creates barrier to particles, micro-organisms, and nonhazardous liquids
- Lowest level particle shedding
- Recyclable
- Serged seams
- Standard hood
- Elastic wrists and ankles
- Zipper closure
- Use in controlled environments with high standards for particle and microbiological contamination control
- Pharmaceutical, biotech, medical device, electronic settings
Specifications
| Cleanroom Coveralls | |
| Unisex | |
| White | |
| Elastic | |
| Zipper | |
| Non-sterile | |
| Yes | |
| 91.4 to 96.5 cm | |
| 24.25 in. | |
| Class 1000-100,000 (ISO 6-8) |
| Medium | |
| Tyvek | |
| Single-use | |
| Elastic | |
| Serged | |
| Tears | |
| 36 to 38 in. | |
| 84.4 cm | |
| Bulk |