Learn More
DuPont™ Tyvek™ IsoClean™ Hooded Coverall with Attached Hood Boots
Clean-processed coveralls provide an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash.
Supplier:  DuPont™ IC105SWH2X00250C
 View more versions of this product
Catalog No. 17-685-208
Only null left
Explore available promotions
Alert:
Should not be worn around open flame, heat source, sparks or ignition sources.
Description
Description
  • Clean-processed garments offer lowest level of particle shedding within DuPont product portfolio
  • Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam
  • Coverall has elastic openings for tighter fit at wrist and ankle
  • Attached thumb loops to minimize bunching of sleeves.
  • Attached hood with elastic around face opening
  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Manufactured under specifications that do not contain natural rubber latex
Specifications
Specifications
Cleanroom Coveralls
Unisex
White
Elastic, Attached Boots
Hooded
Zipper
Non-sterile
43 to 46 in.
109.2 to 116.84 cm
Set
Barrier to Dry Particles, Microorganisms, and Non-Hazardous Liquids in Controlled Environments
97.15 cm
6.1 to 6.5 ft.
Clean-processed
2X-Large
Tyvek
Single-use
Thumb Loop
Elastic
Serged
Elastic wrist and ankle, standard hood with elastic opening, set sleeves, attached thumb loops, attached boot with PVC soles
30.5 in.
77.47 cm
5 mil
38.25 in.
28.75 in.
185.92 to 198.12 cm
Class 1000 (ISO 6)
Product Suggestions
Product Suggestions