missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™ IsoClean™ Hooded Coverall with Attached Hood Boots
Clean-processed coveralls provide an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash.
Supplier: DuPont™ IC105SWHLG00250C
Alert:
Should not be worn around open flame, heat source, sparks or ignition sources.
Description
- Clean-processed garments offer lowest level of particle shedding within DuPont product portfolio
- Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam
- Coverall has elastic openings for tighter fit at wrist and ankle
- Attached thumb loops to minimize bunching of sleeves.
- Attached hood with elastic around face opening
- Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
- Manufactured under specifications that do not contain natural rubber latex
Specifications
| Cleanroom Coveralls | |
| Unisex | |
| White | |
| Elastic, Attached Boots | |
| Hooded | |
| Zipper | |
| Non-sterile | |
| 38 to 40 in. | |
| 96.52 to 101.6 cm | |
| Set | |
| Barrier to Dry Particles, Microorganisms, and Non-Hazardous Liquids in Controlled Environments | |
| 88.9 cm | |
| 5.8 to 6 ft. | |
| Clean-processed |
| Large | |
| Tyvek | |
| Single-use | |
| Thumb Loop | |
| Elastic | |
| Serged | |
| Elastic wrist and ankle, standard hood with elastic opening, set sleeves, attached thumb loops, attached boot with PVC soles | |
| 29 in. | |
| 73.66 cm | |
| 5 mil | |
| 35 in. | |
| 25.75 in. | |
| 176.78 to 182.88 cm | |
| Class 1000 (ISO 6) |