missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ IsoClean™ Series 105 Hooded Coveralls with Attached Boots
GREENER_CHOICE

Catalog No. 17685204
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Height (English):
5.11 to 6.3 ft.
5.5 to 5.10 ft.
5.8 to 6 ft.
6.1 to 6.5 ft.
6.2 to 6.6 ft.
Height (Metric):
155.75 to 192.04 cm
167.64 to 155.44 cm
176.78 to 182.88 cm
185.92 to 198.12 cm
188.97 to 201.16 cm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Height (English) Height (Metric)
17685204 3X-Large 6.2 to 6.6 ft. 188.97 to 201.16 cm
17685200 Medium 5.5 to 5.10 ft. 167.64 to 155.44 cm
17685201 Large 5.8 to 6 ft. 176.78 to 182.88 cm
17685202 X-Large 5.11 to 6.3 ft. 155.75 to 192.04 cm
17685203 2X-Large 6.1 to 6.5 ft. 185.92 to 198.12 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17685204 Supplier DuPont™ Supplier No. IC105SWH3X002500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

IsoClean delivers an ideal balance of protection, durability, and comfort. Bulk packaged in double transparent poly liners.

  • Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam
  • Coverall has elastic openings for tighter fit at wrist and ankle
  • Attached thumb loops to minimize bunching of sleeves
  • Attached hood with elastic around face opening
  • Front zipper closure for easy donning and doffing
  • Attached boots with PVC soles that provide enhanced skid-resistance and durability
  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Manufactured under specifications that do not contain natural rubber latex
  • Bulk packaged, Nonsterile

Specifications

Product Type Cleanroom Coveralls
Size 3X-Large
Gender Unisex
Material Tyvek
Color White
Disposable Single-use
Waist Style Non-elastic
Ankle Style Elastic, Attached Boots
Cuff Style Thumb Loop
Neck Style Hooded
Hood Type Elastic
Closure Type Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Particle, Non-toxic Particulates
Description Elastic wrist and ankle, standard hood with elastic opening, set sleeves, attached thumb loops, attached boot with PVC soles
Size (English) Chest 46 to 52 in.
Size (English) Inseam 31.5 in.
Size (Metric) Chest 116.8 to 132.08 cm
Size (Metric) Inseam 80 cm
Sleeve Style Set
Thickness 5.4 mil
For Use With (Application) Barrier to Dry Particles, Microorganisms, and Non-Hazardous Liquids in Controlled Environments
Length (English) Sleeve 38.5 in.
Length (Metric) Sleeve 97.79 cm
Width (English) Chest 30.25 in.
Height (English) 6.2 to 6.6 ft.
Height (Metric) 188.97 to 201.16 cm
Packaging Bulk
Cleanroom Class Class 1000 (ISO 6)
Show More Show Less