missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™ IsoClean™ Series 105 Hooded Coveralls with Attached Boots
IsoClean delivers an ideal balance of protection, durability, and comfort. Bulk packaged in double transparent poly liners.
Supplier: DuPont™ IC105SWH2X002500
Description
- Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam
- Coverall has elastic openings for tighter fit at wrist and ankle
- Attached thumb loops to minimize bunching of sleeves
- Attached hood with elastic around face opening
- Front zipper closure for easy donning and doffing
- Attached boots with PVC soles that provide enhanced skid-resistance and durability
- Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
- Manufactured under specifications that do not contain natural rubber latex
- Bulk packaged, Nonsterile
Specifications
| Cleanroom Coveralls | |
| Unisex | |
| White | |
| Elastic, Attached Boots | |
| Hooded | |
| Zipper | |
| Non-sterile | |
| 43 to 46 in. | |
| 109.2 to 116.84 cm | |
| Set | |
| Barrier to Dry Particles, Microorganisms, and Non-Hazardous Liquids in Controlled Environments | |
| 97.15 cm | |
| 6.1 to 6.5 ft. | |
| Bulk |
| 2X-Large | |
| Tyvek | |
| Single-use | |
| Thumb Loop | |
| Elastic | |
| Serged | |
| Elastic wrist and ankle, standard hood with elastic opening, set sleeves, attached thumb loops, attached boot with PVC soles | |
| 30.5 in. | |
| 77.47 cm | |
| 5.4 mil | |
| 38.25 in. | |
| 28.75 in. | |
| 185.92 to 198.12 cm | |
| Class 1000 (ISO 6) |