missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ IsoClean™ Series 105 White Coveralls
GREENER_CHOICE

Catalog No. 17685214
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Height (English):
5.11 to 6.3 ft.
5.5 to 5.10 ft.
5.8 to 6 ft.
6.1 to 6.5 ft.
6.2 to 6.6 ft.
Height (Metric):
155.75 to 192.04 cm
167.64 to 155.44 cm
176.78 to 182.88 cm
185.92 to 198.12 cm
188.97 to 201.16 cm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Height (English) Height (Metric)
17685214 3X-Large 6.2 to 6.6 ft. 188.97 to 201.16 cm
17685210 Medium 5.5 to 5.10 ft. 167.64 to 155.44 cm
17685211 Large 5.8 to 6 ft. 176.78 to 182.88 cm
17685212 X-Large 5.11 to 6.3 ft. 155.75 to 192.04 cm
17685213 2X-Large 6.1 to 6.5 ft. 185.92 to 198.12 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17685214 Supplier DuPont™ Supplier No. IC105SWH3X0025CS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ideal for use in controlled environments to protect against particulates and other contaminants.

  • Light and durable coveralls provide coverage, including feet, while staying comfortable.
  • Use in controlled environments with high standards for particle and microbiological contamination control.
  • Clean processed and sterile
  • Spinning process creates barrier to particles, micro-organisms, and non-hazardous liquids
  • Lightweight, comfortable and durable
  • Lowest level particle shedding
  • Recyclable
  • Serged seams
  • Standard hood, elastic opening
  • Elastic wrists and ankles
  • Attached thumb loops
  • Zipper closure
  • Attached boots, PVC sole
  • Pharmaceutical, biotech, medical device, electronic settings

Specifications

Product Type Cleanroom Coveralls
Size 3X-Large
Gender Unisex
Material Tyvek
Color White
Disposable Single-use
Waist Style Non-elastic
Ankle Style Elastic, Attached Boots
Cuff Style Thumb Loop
Neck Style Hooded
Hood Type Elastic
Closure Type Zipper
Seam Type Serged
Sterility Sterile
Resistance Particle, Non-toxic Particulates
Description Elastic wrist and ankle, attached thumb loops, standard hood with elastic opening, attached boot with PVC soles, set sleeves
Model IC105SWH
Size (English) Chest 46 to 52 in.
Size (English) Inseam 31.5 in.
Size (Metric) Chest 116.8 to 132.08 cm
Size (Metric) Inseam 80 cm
Thickness 5 mil
For Use With (Application) Barrier to dry particles, microorganisms, and non-hazardous liquids
Length (English) Sleeve 38.5 in.
Length (Metric) Sleeve 97.79 cm
Height (English) 6.2 to 6.6 ft.
Height (Metric) 188.97 to 201.16 cm
Cleanroom Class Class 100-100,000 (ISO 5-8)
Show More Show Less