missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ IsoClean™ Serged Seam Cleanroom Frocks
GREENER_CHOICE

Catalog No. 17999000
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Sterility:
Non-sterile
Sterile
Packaging Type:
Bulk Packaged
Individually Packaged
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Sterility Packaging Type
17999000 4X-Large Non-sterile Bulk Packaged
17988072 3X-Large Non-sterile Bulk Packaged
17988066 3X-Large Sterile Individually Packaged
17988065 2X-Large Sterile Individually Packaged
17988071 2X-Large Non-sterile Bulk Packaged
17988070 X-Large Non-sterile Bulk Packaged
17988064 X-Large Sterile Individually Packaged
17988069 Large Non-sterile Bulk Packaged
17988063 Large Sterile Individually Packaged
17988068 Medium Non-sterile Bulk Packaged
17988062 Medium Sterile Individually Packaged
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Garments made of Tyvek™ should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Catalog No. 17999000 Supplier DuPont™ Supplier No. IC262SWH4X00300B

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Garments made of Tyvek™ should not be used around heat, flames, sparks or in potentially flammable or explosive environments.

Available in sterile or non-sterile versions, Tyvek™ cleanroom frocks provide an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash in ISO 5 (Class 100) and above environments.

  • Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam.
  • Elastic opening for tighter fit at wrist
  • Mandarin collar with snap for secure fit
  • Body-length jacket design with front snap closures for easy donning and doffing
  • Raglan sleeve design for greater range of motion and comfort.
  • Generous sizing
  • Non-sterile versions are bulk packaged in double transparent poly liners
  • Sterile versions are individually packaged
  • Manufactured under specifications that do not contain natural rubber latex
  • Sterile frocks are gamma sterilized to SAL of 10-6

Specifications

Size 4X-Large
Material Tyvek
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Snaps
Cuff Style Elastic
Collar Type High Mandarin
Sleeve Style Raglan Sleeve
Packaging Type Bulk Packaged
Cleanroom Class Class 100 (ISO 5)
Product Type Controlled Environments Frock
Seam Type Serged
For Use With (Application) Cleanroom, Pharmaceutical, Biotechnology, Medical Device Manufacturing, and Laboratory Environments
Show More Show Less