missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™ IsoClean™ Serged Seam Cleanroom Frocks
Available in sterile or non-sterile versions, Tyvek™ cleanroom frocks provide an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash in ISO 5 (Class 100) and above environments.
Supplier: DuPont™ IC262SWHXL00300B
Alert:
Garments made of Tyvek™ should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Description
- Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam.
- Elastic opening for tighter fit at wrist
- Mandarin collar with snap for secure fit
- Body-length jacket design with front snap closures for easy donning and doffing
- Raglan sleeve design for greater range of motion and comfort.
- Generous sizing
- Non-sterile versions are bulk packaged in double transparent poly liners
- Sterile versions are individually packaged
- Manufactured under specifications that do not contain natural rubber latex
- Sterile frocks are gamma sterilized to SAL of 10-6
Specifications
| Controlled Environments Frock | |
| Tyvek | |
| Single-use | |
| Snaps | |
| Serged | |
| 38 to 40 in. | |
| Bulk Packaged | |
| 121.28 cm | |
| 101.6 cm |
| X-Large | |
| White | |
| Non-sterile | |
| Elastic | |
| High Mandarin | |
| Raglan Sleeve | |
| 47.75 in. | |
| 40 in. | |
| Class 100 (ISO 5) |