Learn More
DuPont™ Tyvek™ IsoClean™ Nonsterile Bouffants
GREENER_CHOICE
Catalog No. 17988116
Change view
Click to view available options
Packaging Type:
Bulk Packed
Clean Packed
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Type
17988116 Clean Packed
17988115 Bulk Packed
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Catalog No. 17988116 Supplier DuPont™ Supplier No. IC729SWH0002500C
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Creates a barrier to dry particles, microorganisms, and nonhazardous liquids. Suitable for ISO 5 (class 100) or higher cleanroom environments.
  • Serged seams
  • Elastic headband
  • 21.5 in. dia.
  • Comfortable, lightweight and durable
  • White
Specifications
Product Type Cleanroom Bouffant
Size 21 in.
Material Tyvek
Color White
Disposable Single-use
Model IC729SWH0002500C
Seam Type Serged
Certifications/Compliance Category III PPE
Packaging Type Clean Packed
Quantity 250/Cs.
Diameter (English) 21.5 in.
Includes Elastic Headband
Cleanroom Class Class 10 (ISO 4)
Show More Show Less