missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™ IsoClean™ Boot Covers
Provides inherent barrier to dry particles, microorganisms, and non-hazardous liquids. Available in bulk or clean-processed packaging.
Supplier: DuPont™ IC458BWHXL01000C
Description
- Made from flashspun polyolefin protective material using proprietary flash spinning process
- Comfortable, lightweight and durable
- Ankle ties for secure fit
- Covered elastic opening
- Available with choice of seams, openings and soles
- Height: 18 in. (45.7cm)
- White
IC457S
- Serged seams with multiple interlocking threads sewn around raw edges of garment to create strong, stress-resistant seam
- With PVC soles that provide skid-resistance and durability
- Available bulk packaged in double transparent poly liners
- Bound seams covered with garment fabric to reinforce seam and to reduce potential for particle penetration
- With Gripper™ soles for enhanced skid-resistance and durability
- Available clean-processed
- Available bulk packaged in double transparent poly liners
Specifications
| X-Large | |
| Yes | |
| Non-sterile | |
| Anti-skid | |
| 5 mil | |
| Clean Processed | |
| 41.91 cm | |
| 46.99 cm |
| Tyvek | |
| White | |
| Elastic Top with Ankle Ties | |
| Bound | |
| 16 CFR 1610 Class 1 | |
| 16.5 in. | |
| 18.5 in. | |
| Class 1000 (ISO 6) |