missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ IsoClean™ Boot Covers

Catalog No. 17988088
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
Seam Type:
Bound
Serged
Packaging Type:
Bulk Packed
Clean Processed
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Seam Type Packaging Type
17988088 Small Serged Bulk Packed
17988089 Medium Serged Bulk Packed
17988090 Large Serged Bulk Packed
17988091 X-Large Serged Bulk Packed
17988100 Medium Bound Bulk Packed
17988101 Large Bound Bulk Packed
17988102 X-Large Bound Bulk Packed
19100407 Medium Bound Clean Processed
17988096 Large Bound Clean Processed
17988097 X-Large Bound Clean Processed
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 17988088 Supplier DuPont™ Supplier No. IC457SWHSM01000B

Provides inherent barrier to dry particles, microorganisms, and non-hazardous liquids. Available in bulk or clean-processed packaging.

  • Made from flashspun polyolefin protective material using proprietary flash spinning process
  • Comfortable, lightweight and durable
  • Ankle ties for secure fit
  • Covered elastic opening
  • Available with choice of seams, openings and soles
  • Height: 18 in. (45.7cm)
  • White

IC457S

  • Serged seams with multiple interlocking threads sewn around raw edges of garment to create strong, stress-resistant seam
  • With PVC soles that provide skid-resistance and durability
  • Available bulk packaged in double transparent poly liners
IC458S
  • Bound seams covered with garment fabric to reinforce seam and to reduce potential for particle penetration
  • With Gripper™ soles for enhanced skid-resistance and durability
  • Available clean-processed
  • Available bulk packaged in double transparent poly liners

Specifications

Size Small
Material Tyvek
Latex Free Yes
Color White
Sterility Non-sterile
Closure Type Elastic Top with Ankle Ties
Sole Type Anti-skid
Seam Type Serged
Thickness 5.4 mil
Certifications/Compliance 16 CFR 1610 Class 1
Packaging Type Bulk Packed
Length (English) 10.5 in.
Length (Metric) 26.67 cm
Height (English) 18.5 in.
Height (Metric) 46.99 cm
Cleanroom Class Class 1000 (ISO 6)
Show More Show Less