missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ DuPont™ Tyvek™ IsoClean™ Boot Cover, IC447S - Option CS
Description
Tyvek™ IsoClean™ delivers an ideal balance of protection, durability and comfort. Made using a patented flash spinning process, Tyvek™ provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Designed for controlled environments with excellent barrier to particles, microorganisms, and non-hazardous liquids. Suitable for cleanroom applications in pharmaceutical, biotech, medical device manufacturing, and laboratory settings.
- Garments are gamma sterilized to SAL of 10-6.
- Clean-processed garments offer lowest level of particle shedding within DuPont product portfolio
- Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam
- Elastic at ankles for secure fit Gripper™ soles provide enhanced skid-resistance and durability
- 18 in. high boot cover with elastic top provides coverage up to calf
Specifications
Specifications
| Size | Small |
| Material | Tyvek IsoClean |
| Latex Free | No |
| Color | White |
| Disposable | Single-use |
| Sterility | Sterile |
| Closure Type | Elastic Top and Ankles |
| Sole Type | Skid-resistant |
| Resistance | Abrasion |
| Product Type | Controlled Environments Boot Covers |
| Show More |