missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ IsoClean™ Frock
GREENER_CHOICE

Catalog No. 17990519
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17990519 4X-Large
17990513 Small
17990514 Medium
17990515 Large
17990516 X-Large
17990517 2X-Large
17990518 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Should not be worn around open flame, heat source, sparks or ignition sources.
Catalog No. 17990519 Supplier DuPont™ Supplier No. IC264SWH4X0030CS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Should not be worn around open flame, heat source, sparks or ignition sources.

Sterile and clean-processed frocks offer lowest level of particle shedding within the DuPont portfolio. Tyvek™ provides an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash.

  • Frocks are gamma sterilized to SAL of 10-6
  • Clean-processed
  • Serged seams have multiple interlocking threads sewn around the raw edges
  • Elastic openings for tighter fit at wrist
  • Bound neck for lower particle shedding
  • Front zipper closure for easy donning and doffing
  • Raglan sleeve design for greater range of motion and comfort
  • A-line silhouette
  • Individually packaged, 30 per case

Specifications

Size 4X-Large
Material Tyvek
Color White
Disposable Single-use
Sterility Sterile
Closure Type Zipper
Cuff Style Elastic
Collar Type Bound
Style IsoClean™ 264 Series
Sleeve Style Raglan Sleeve
Packaging Type Clean-processed, Individually Packaged
Length (Metric) 144.8 cm
Length (English) Sleeve 46 in.
Length (Metric) Sleeve 116.8 cm
Length (English) 57 in.
Product Type Controlled Environments Frock
Seam Type Serged
For Use With (Application) Cleanroom, Pharmaceutical, Biotechnology, Medical Device Manufacturing, and Laboratory Environments
Show More Show Less