Learn More
DuPont™ Tyvek™ IsoClean™ Frock
Sterile and clean-processed frocks offer lowest level of particle shedding within the DuPont portfolio. Tyvek™ provides an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash.
Supplier:  DuPont™ IC264SWH2X0030CS
 Afficher plus de versions de ce produit
Numéro de catalogue. 17-990-517
Il en reste null
Découvrez plus d'offres spéciales
Avertissement:
Should not be worn around open flame, heat source, sparks or ignition sources.
Description
Description
  • Frocks are gamma sterilized to SAL of 10-6
  • Clean-processed
  • Serged seams have multiple interlocking threads sewn around the raw edges
  • Elastic openings for tighter fit at wrist
  • Bound neck for lower particle shedding
  • Front zipper closure for easy donning and doffing
  • Raglan sleeve design for greater range of motion and comfort
  • A-line silhouette
  • Individually packaged, 30 per case
Spécifications
Spécifications
Controlled Environments Frock
Tyvek
Single-use
Zipper
Serged
IsoClean™ 264 Series
Clean-processed, Individually Packaged
132.1 cm
108.5 cm
2X-Large
White
Sterile
Elastic
Bound
Raglan Sleeve
53 in.
42.75 in.
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS