Learn More
Keystone Cleanroom Products DuPont™ Tyvek™ Autoclave Pouch with Steam Indicator Dot
Manufactured in a ISO6 cleanroom to ensure cleanliness.
Supplier:  Keystone Cleanroom Products COVTVKHS12X32
 View more versions of this product
Catalog No. 19-061-315
Only null left
Explore available promotions
Description
Description
  • Designed for steam sterilization of pharmaceutical production equipment and components
  • 1073B Tyvek™ construction allows for optimal steam penetration during sterilization
  • Excellent impact resistance
  • Consistently strong factory seals
  • Exceptional microbial barrier
  • Resists tearing and punctures from sharp corners and heavy machine parts
  • Processed and packaged in a controlled cleanroom environment
  • Lot traceable
Specifications
Specifications
Autoclavable
Certificate of Conformance (COC)
White
Steam Sterilization
Tyvek
Yes
100/Cs.
32 in. x 12 in.
81 cm
30.5 cm
Application Dependent
Heat Seal
Single-use
Pharmaceutical Equipment Change Parts/Components (Stoppers, Seals, etc.)
Non-sterile
1615 g/m2/24 hr
Steam
32 in.
12 in.
Autoclave Bag
Product Suggestions
Product Suggestions