Learn More
DuPont™ Tyvek™ 400 Disposable Pants
GREENER_CHOICE
Catalog No. 19067055
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
7X-Large
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19067055 Small
0136120B Medium
19067054 Large
0136120E 2X-Large
0136120F 3X-Large
0136120D X-Large
0136120G 4X-Large
17500293 5X-Large
17500294 6X-Large
17500295 7X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 19067055 Supplier DuPont™ Supplier No. TY350SWHSM005000
Designed disposable pants featuring lightweight, chemically and biologically inert, tear-resistant
  • Pants with open ankle and elastic at waist for easy donning and removal
  • Suitable for applications that do not require the full body coverage of a coverall
  • Typically worn with TY303S WH shirt
  • Manufactured under specifications that do not contain natural rubber latex
Specifications
Size Small
Product Type General Purpose Pants
Gender Unisex
Material Tyvek
Color White
Ankle Style Open
Seam Type Serged
No. of Pockets None
Size (English) Waist 22 in.
Waist Style Elastic