Learn More
DuPont™ Tyvek™ 400 Disposable Pants
Designed disposable pants featuring lightweight, chemically and biologically inert, tear-resistant
Supplier:  DuPont™ TY350SWHMD005000
 View more versions of this product
Catalog No. 01-361-20B
Only null left
Explore available promotions
Description
Description
  • Pants with open ankle and elastic at waist for easy donning and removal
  • Suitable for applications that do not require the full body coverage of a coverall
  • Typically worn with TY303S WH shirt
  • Manufactured under specifications that do not contain natural rubber latex
Specifications
Specifications
Single-use Pants
Unisex
White
Serged
23.5 in.
Lead and Asbestos Abatement/Remedation, General Maintenance/Operations, Spray Painting, General Clean-up
Medium
Tyvek
Open
69.85 cm
59.69 cm
Product Suggestions
Product Suggestions