Learn More
DuPont™ Tychem™ 10000 Training Suit
Catalog No. p-3775783
Change view
Click to view available options
Size:
Medium
Large
2X-Large
3X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19715357 2X-Large
19715358 3X-Large
19715354 Medium
19715355 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19715357 Supplier DuPont™ Supplier No. TK587SLY2X000100
Only null left
Tychem 10000 fully encapsulated training suits offer an economical and effective option for HazMat training
  • Tychem™ 10000 fully encapsulated training suits offer an economical and effective option for HazMat training. Same fabric and garment design as standard Tychem™ 10000 suits.
  • Multiple interlocking threads are sewn around the raw edges of garment material to create a strong, stress-resistant seam
  • Extra wide (EX) single layer faceshield (20 mil PVC) for wide viewing angle
  • Rear entry design keeps closure away from direct contact with frontal chemical exposure and provides ability to change air bottle on SCBA without removing the entire garment
  • Integrated butyl gloves
  • Internal adjustable waist belt system for support and improved fit
  • Clearly labeled for training use only
  • Manufactured under specifications that do not contain natural rubber latex
  • Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant
Specifications
Color Lime Yellow
Material Tychem
Size 2X-Large
Ankle Style Boot Overflaps
Closure Type Zipper
Seam Type Serged