Learn More
DuPont™ Tychem™ 10000 Training Suit
Catalog No. 19715354
Change view
Click to view available options
Size:
Medium
Large
2X-Large
3X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19715354 Medium
19715357 2X-Large
19715358 3X-Large
19715355 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19715354 Supplier DuPont™ Supplier No. TK587SLYMD000100
Only null left
Tychem 10000 fully encapsulated training suits offer an economical and effective option for HazMat training
  • Tychem™ 10000 fully encapsulated training suits offer an economical and effective option for HazMat training. Same fabric and garment design as standard Tychem™ 10000 suits.
  • Multiple interlocking threads are sewn around the raw edges of garment material to create a strong, stress-resistant seam
  • Extra wide (EX) single layer faceshield (20 mil PVC) for wide viewing angle
  • Rear entry design keeps closure away from direct contact with frontal chemical exposure and provides ability to change air bottle on SCBA without removing the entire garment
  • Integrated butyl gloves
  • Internal adjustable waist belt system for support and improved fit
  • Clearly labeled for training use only
  • Manufactured under specifications that do not contain natural rubber latex
  • Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant
Specifications
Color Lime Yellow
Material Tychem
Size Medium
Ankle Style Boot Overflaps
Closure Type Zipper
Seam Type Serged