missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tychem™ 4000 Hooded Coverall

Catalog No. 19822502
Change view
Click to view available options
Size:
2X-Large
3X-Large
Height (English):
6 to 6.4 ft.
6.2 to 6.4 ft.
Height (Metric):
1.82 to 1.95 m
1.88 to 1.95 m
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Height (English) Height (Metric)
19822502 2X-Large 6 to 6.4 ft. 1.82 to 1.95 m
19822503 3X-Large 6.2 to 6.4 ft. 1.88 to 1.95 m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Garments made of Tyvek™ should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Catalog No. 19822502 Supplier DuPont™ Supplier No. SL128TWH2X000600
Garments made of Tyvek™ should not be used around heat, flames, sparks or in potentially flammable or explosive environments.

Protects against a range of chemicals in a variety of industries

  • Zipper features a semi-automatic locking slider allowing the puller to stay down and in a locked position
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.
  • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
  • Attached respirator-fit hood with elastic around face opening, designed to cover neck and chin and fit around respirator face mask
  • Strong zipper for tough, flexible closure
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
  • Integrated socks composed of garment material
  • Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion
  • Manufactured under specifications that do not contain natural rubber latex

Order Info

19-130-1 Seriesis manufactured in the U.S.

†Attached socks are to be worn inside the user's own chemical protective boots. The boot splash cover adds additional protection. These attached socks do not have adequate durability or slip resistance to be worn as outer footwear covering.

Specifications

Size 2X-Large
Gender Unisex
Material Tychem
Color White
Disposable Single-use
Ankle Style Elastic, Attached Socks
Cuff Style Elastic
Hood Type Attached, Elastic Face
Closure Type Zipper
Seam Type Taped
Certifications/Compliance ASTM D1777
Description Saranex™ 23-P Film Laminated to Tyvek™ Fabric; Elastic around the face, elastic wrist, zipper shall be covered with storm flap with adhesive closure
Size (English) Chest 44.75 to 48.25 in.
Size (English) Inseam 31.25 in.
Size (Metric) Chest 113.66 to 122.55 cm
Size (Metric) Inseam 79.37 cm
Thickness 0.3 mm
For Use With (Application) Chemical mixing, hazardous remediation, emergency medical response, paint spraying and nuclear environments
Length (English) Sleeve 38.5 in.
Length (Metric) Sleeve 97.79 cm
Width (English) Chest 29 in.
Height (English) 6 to 6.4 ft.
Height (Metric) 1.82 to 1.95 m
Weight (English) 0.22 lb.
Weight (Metric) 0.1 kg
Show More Show Less