missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tychem™ 4000 Coverall (all)

Catalog No. 19039003F
Change view
Click to view available options
Size:
X-Large
3X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19039003F 3X-Large
19039003D X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19039003F Supplier DuPont™ Supplier No. SL125TWH3X000600
  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
  • Laydown collar
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
  • Elastic opening for tighter fit at ankle
  • Manufactured under specifications that do not contain natural rubber latex
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.

Specifications

Size 3X-Large
Gender Unisex
Material Tychem
Color White
Waist Style Waist Ties
Ankle Style Elastic
Cuff Style Elastic
Closure Type Zipper
Seam Type Taped
Description Elastic wrist, 44 in. long, neck loop with snap and waist ties
Size (English) Chest 44.75 to 48.25 in.
Size (Metric) Chest 113.6 to 122.55 cm
Sleeve Style Long sleeve
Thickness 12 mil
For Use With (Application) Chemical mixing, hazardous remediation, emergency medical response, paint spraying and nuclear environments
Length (English) 46.5 in.
Length (English) Sleeve 33.75 in.
Length (Metric) 2.906 cm
Length (Metric) Sleeve 85.72 cm
Height (English) 6.2 to 6.4 ft.
Height (Metric) 1.88 to 1.95 m
Show More Show Less