missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tychem™ 4000 Sleeved Apron

Catalog No. 19162789
Change view
Click to view available options
Size:
Small
4X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19162789 Small
19162788 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19162789 Supplier DuPont™ Supplier No. SL275TWHSM002500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

DuPont Tychem 4000 Sleeved Apron. Elastic Wrists. 44 in. Long. Neck Loop w/ Snaps and Waist Ties. Taped Seams.

  • Knee-length apron/gown design with attached sleeves
  • Available in multiple sizes
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.
  • Provides enhanced frontal protection
  • Taped seams provide strong chemical resistance against heavy liquid splashes
  • A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing
  • Open back for wearer comfort.
  • Bound seams around neck opening with snap closure
  • Ties at waist
  • Elastic opening for tighter fit at wrist
  • 44 in. long for neck-to-knee coverage
  • Manufactured under specifications that do not contain natural rubber latex

Specifications

Type Chemical-resistant Apron
Size Small
Material Tychem
Color White
Disposable Single-use
Closure Type Neck Snap, Waist Ties
Sterility Non-sterile
For Use With (Application) Waste management, hazardous response and nuclear environments
Length (English) 44 in.
Length (Metric) 111.7 cm
Seam Type Taped
Show More Show Less