missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tychem™ 4000 Sleeved Apron

Catalog No. 19162788
Change view
Click to view available options
Size:
Small
4X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19162788 4X-Large
19162789 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19162788 Supplier DuPont™ Supplier No. SL275TWH4X002500

DuPont Tychem 4000 Sleeved Apron. Elastic Wrists. 44 in. Long. Neck Loop w/ Snaps and Waist Ties. Taped Seams.

  • Knee-length apron/gown design with attached sleeves
  • Available in multiple sizes
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.
  • Provides enhanced frontal protection
  • Taped seams provide strong chemical resistance against heavy liquid splashes
  • A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing
  • Open back for wearer comfort.
  • Bound seams around neck opening with snap closure
  • Ties at waist
  • Elastic opening for tighter fit at wrist
  • 44 in. long for neck-to-knee coverage
  • Manufactured under specifications that do not contain natural rubber latex

Specifications

Size 4X-Large
Material Tychem
Color White
Closure Type Waist Ties
Includes Attached Long Sleeves
For Use With (Application) Waste management, hazardous response and nuclear environments
Length (English) 44 in.
Length (Metric) 111.7 cm
Seam Type Taped