missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tychem™ 4000 Hooded Coverall

Catalog No. 19162781
Change view
Click to view available options
Size:
5X-Large
6X-Large
7X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19162781 7X-Large
19162779 5X-Large
19162780 6X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19162781 Supplier DuPont™ Supplier No. SL127TWH7X000600
  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
  • Attached hood with elastic around face opening.
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
  • Elastic opening for tighter fit at ankle
  • Manufactured under specifications that do not contain natural rubber latex
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.

Specifications

Size 7X-Large
Gender Unisex
Material Tychem
Color White
Ankle Style Elastic
Neck Style Hooded
Hood Type Attached
Closure Type Front Zipper
Seam Type Taped
Description Attached hood
For Use With (Application) Waste management, hazardous response and nuclear,environments
Show More Show Less