missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tychem™ 4000 Coverall (all)

Catalog No. 19813434
Change view
Click to view available options
Size:
Medium
X-Large
3X-Large
Height (English):
5.3 to 5.7 ft.
6.2 to 6.4 ft.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Height (English)
19813434 Medium 5.3 to 5.7 ft.
19813438 3X-Large 6.2 to 6.4 ft.
19813436 X-Large -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19813434 Supplier DuPont™ Supplier No. SL121TWHMD000400
Only null left

Tychem™ 4000 Coverall includes elastic wrists and attached socks.

  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
  • Laydown collar
  • Elastic opening for tighter fit at wrist
  • Integrated socks composed of garment material
  • Manufactured under specifications that do not contain natural rubber latex
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.

Specifications

Size Medium
Gender Unisex
Material Tychem
Color White
Disposable Single-use
Ankle Style Attached Socks
Closure Type Zipper
Seam Type Taped
Certifications/Compliance ASTM D1777, ASTM D3776, ASTM D3786, ASTM D5034, 16 CFR 1610
Description Storm flap with tape closure; Multi-layer film barrier laminated
Size (English) Chest 35.25 to 38.75 in.
Size (English) Inseam 28 in.
Size (Metric) Chest 89.53 to 98.42 cm
For Use With (Application) Chemical mixing, hazardous remediation, emergency medical response, paint spraying, and nuclear environments.
Length (English) Sleeve 33.75 in.
Length (Metric) Sleeve 85.72 cm
Width (English) Chest 24.25 in.
Width (Metric) Chest 61.59 cm
Height (English) 5.3 to 5.7 ft.
Height (Metric) 161.54 to 173.73 cm
Show More Show Less