missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tychem™ 5000 Hooded Coverall

Catalog No. p-2889507
Change view
Click to view available options
Collar Type:
3-panel, Respirator Fit
Standard Collar
Size:
Medium
Large
X-Large
2X-Large
3X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Collar Type Size
191301023B 3-panel, Respirator Fit Medium
191301023C 3-panel, Respirator Fit Large
191301023E 3-panel, Respirator Fit 2X-Large
191301023D 3-panel, Respirator Fit X-Large
191301024C Standard Collar Large
191301023F 3-panel, Respirator Fit 3X-Large
191301024D Standard Collar X-Large
191301024F Standard Collar 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 191301023B Supplier DuPont™ Supplier No. C3185TTNMD000600
Only null left

Integrated gloves eliminate need for taping and provide improved protection

  • Zipper features a semi-automatic locking slider allowing the puller to stay down and in a locked position
  • Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion
  • Manufactured under specifications that do not contain natural rubber latex
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.
  • Tychem™ CPF3 HD garments were designed with extensive input from first responders to ensure that HAZMAT, law enforcement, military and hospital personnel get the protection they need from chemical warfare agents (including Sarin, Mustard, and Lewisite) and toxic industrial chemicals (TIC's).
  • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
  • Three-piece hood design with longer zipper that extends to chin for complete coverage of neck area and good fit around a respirator
  • Larger zipper pull is easier to grasp for easier donning and doffing
  • Storm flap covers zipper which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper
  • Attached internal gloves composed of five layer laminate film for a broad range of chemical protection
  • Neoprene outer gloves deliver high tactility and dexterity to handle tools or weapons
  • Integrated socks composed of garment material

Order Info

Basis Weight: 4.5 oz./yd.2 per ASTM™ D751. Ball Burst (md/cd): 47lbf per ASTM D750. Grab Tensile Strength (md/cd): 60/50 ft.-lb per ASTM D751.

†Review separate permeation data on both gloves and the fabric before selecting an ensemble.

‡Attached socks are to be worn inside the user's own chemical protective boots. The boot splash cover adds protection. These attached socks do not have adequate durability or slip resistance to be worn as outer footwear covering.

Specifications

Size Medium
Gender Unisex
Material Tychem
Color Tan
Ankle Style Attached Socks, Boot Overflaps
Cuff Style Attached Gloves
Neck Style Hooded
Hood Type Respirator Fit
Closure Type Zipper
Seam Type Taped
Collar Type 3-panel, Respirator Fit
Description Respirator fit hood, attached dual layer gloves, attached socks with outer boot flaps; Multilayer film barrier laminated to Polypropylene substrate
Size (English) Chest 34.75 to 38.25 in.
Size (English) Inseam 28 in.
Size (Metric) Chest 88.26 to 97.15 cm
Size (Metric) Inseam 71.12 cm
For Use With (Application) Chemical handling, petrochemical, hazardous materials, waste cleanup, fire departments, industrial hazmat teams, utilities and domestic preparedness
Length (English) Sleeve 35.25 in.
Length (Metric) Sleeve 89.53 cm
Width (English) Chest 24 in.
Height (English) 5.3 to 5.7 ft.
Height (Metric) 1.6 to 1.73 m
Show More Show Less