missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tychem™ 5000 Hooded Coverall

Catalog No. 190881230E
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Certifications/Compliance:
ANSI/ISEA 101-1996, CAN/CGSB-155.20-2000, NFPA 701 (1989), NFPA 99
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Certifications/Compliance
190881230E 2X-Large ANSI/ISEA 101-1996, CAN/CGSB-155.20-2000, NFPA 701 (1989), NFPA 99
190881230G 4X-Large ANSI/ISEA 101-1996, CAN/CGSB-155.20-2000, NFPA 701 (1989), NFPA 99
2705570 Large ANSI/ISEA 101-1996, CAN/CGSB-155.20-2000, NFPA 701 (1989), NFPA 99
2705569 Medium ANSI/ISEA 101-1996, CAN/CGSB-155.20-2000, NFPA 701 (1989), NFPA 99
190881230D X-Large ANSI/ISEA 101-1996, CAN/CGSB-155.20-2000, NFPA 701 (1989), NFPA 99
190881230F 3X-Large ANSI/ISEA 101-1996, CAN/CGSB-155.20-2000, NFPA 701 (1989), NFPA 99
17990507 5X-Large ANSI/ISEA 101-1996, CAN/CGSB-155.20-2000, NFPA 701 (1989), NFPA 99
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Do not use Tychem CPF garments for fire protection. Avoid exposure to open flame or intense heat. Attached socks are to be worn inside the user's own chemical protective boots. The boot splash cover adds protection. These attached socks do not have adequate durability or slip resistance to be worn as outer footwear covering.
Catalog No. 190881230E Supplier DuPont™ Supplier No. C3122TTN2X000600
Do not use Tychem CPF garments for fire protection. Avoid exposure to open flame or intense heat. Attached socks are to be worn inside the user's own chemical protective boots. The boot splash cover adds protection. These attached socks do not have adequate durability or slip resistance to be worn as outer footwear covering.

Protection against chemical splash as well as potential exposure to chemical warfare agents

  • Zipper features a semi-automatic locking slider allowing the puller to stay down and in a locked position
  • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
  • Attached hood with elastic around face opening.
  • Strong zipper for tough, flexible closure
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
  • Integrated socks composed of garment material
  • Manufactured under specifications that do not contain natural rubber latex
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.

Compliance

Meets ANSI™/ISEA™ 101-1996 standards; complies with CAN/CGSB-155.20-2000 for flame resistance; NFPA 701 Standard Methods for Fire Tests for Flame Propagation of Textiles and Films (1989); NFPA 99 Health Care Facilities requirements.

†Attached socks are to be worn inside the user's own chemical protective boots. The boot splash cover adds additional protection. These attached socks do not have adequate durability or slip resistance to be worn as outer footwear covering.

Specifications

Size 2X-Large
Gender Unisex
Material Tychem
Color Tan
Disposable Single-use
Ankle Style Attached Socks
Cuff Style Elastic
Neck Style Hooded
Hood Type Attached, Elastic Face
Closure Type Zipper
Seam Type Taped
Certifications/Compliance ANSI/ISEA 101-1996, CAN/CGSB-155.20-2000, NFPA 701 (1989), NFPA 99
Description Tychem™ CPF 3 a multilayer composite barrier film laminated to a durable 2 oz. Polypropylene substrate
Size (English) Inseam 30 in.
Size (Metric) Chest 115.57 to 124.46 cm
Size (Metric) Waist 147.32 cm
Thickness 0.45 mm
For Use With (Application) Chemical handling, petrochemical, hazardous materials, waste cleanup, fire departments, industrial hazmat teams, utilities and domestic preparedness
Length (English) Sleeve 37 in.
Length (Metric) Sleeve 93.98 cm
Width (English) Chest 29.37 in.
Height (English) 6 to 6.4 ft.
Height (Metric) 1.82 to 1.95 m
Weight (English) 0.27 lb.
Weight (Metric) 0.12 kg
Show More Show Less