missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tychem™ 5000 Hooded Coverall

Catalog No. 19067013
Change view
Click to view available options
Size:
3X-Large
4X-Large
5X-Large
Width (English) Chest:
31.25 in.
33.5 in.
36 in.
Height (English):
6 ft. 4 in. to 6 ft. 7 in.
6 ft. 7 in. to 6 ft. 10 in.
6 ft.2 in. to 6 ft. 4 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Width (English) Chest Height (English)
19067013 5X-Large 36 in. 6 ft. 7 in. to 6 ft. 10 in.
19067011 3X-Large 31.25 in. 6 ft.2 in. to 6 ft. 4 in.
19067012 4X-Large 33.5 in. 6 ft. 4 in. to 6 ft. 7 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19067013 Supplier DuPont™ Supplier No. C3127TTN5X000600

Taped seams provide strong chemical resistance against heavy liquid splashes

  • Zipper features a semi-automatic locking slider allowing the puller to stay down and in a locked position
  • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
  • Attached hood with elastic around face opening.
  • Strong zipper for tough, flexible closure
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
  • Elastic opening for tighter fit at ankle
  • Manufactured under specifications that do not contain natural rubber latex
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.

Specifications

Size 5X-Large
Gender Unisex
Material Tychem
Color Tan
Ankle Style Elastic
Closure Type Zipper
Seam Type Taped
Width (English) Chest 36 in.
Height (English) 6 ft. 7 in. to 6 ft. 10 in.