Learn More
DuPont™ Tychem™ 5000 Coverall (all)
Taped seams provide strong chemical resistance against heavy liquid splashes
Supplier:  DuPont™ C3125TTN4X000600
 View more versions of this product
Catalog No. 19-067-007
Only null left
Explore available promotions
Description
Description
  • Zipper features a semi-automatic locking slider allowing the puller to stay down and in a locked position
  • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
  • Laydown collar
  • Strong zipper for tough, flexible closure
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
  • Elastic opening for tighter fit at ankle
  • Manufactured under specifications that do not contain natural rubber latex
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.
Specifications
Specifications
4X-Large
Tychem
Elastic
Taped
6 ft. 4 in. to 6 ft. 7 in.
Unisex
Tan
Zipper
34 in.
Product Suggestions
Product Suggestions