missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tychem™ CPF 3 Combo Suit
Supplier: DuPont™ C3750TTN2X000600
Description
Tychem 5000 fabric is composed of a multi-layer film barrier laminated to a durable 2.0 oz/yd2 polypropylene substrate. Tychem 5000 fabric is strong and durable for rigorous activities and rugged situations involving liquid splash and provides barrier to a broad range of chemicals. Typical Applications: chemical handling, petro-chemical, hazardous materials/waste clean-up, fire departments, industrial hazmat teams, utilities, and domestic preparedness. Commonly used in domestic preparedness for situations where the potential to exposure to chemicals exist.Jacket and bib overall combination ensemble. Components also sold separatelyComes with a jam fit cuff and glove insert for user supplied glovesSpecifications
| ASTM D751, ASTM D1117, ASTM D3787, ASTM D751, ASTM D2582, ASTM D751, 16CFR1610 | |
| Chemical handling, petro-chemical, hazardous materials/waste clean-up, fire departments, industrial hazmat teams, utilities, and domestic preparedness | |
| Multi-layer Barrier Laminate Film | |
| Open | |
| Elastic | |
| Single-use | |
| 182.88 to 195 cm | |
| 30.7 in. | |
| 73.6 cm | |
| Taped | |
| 71.7 cm |
| Tan | |
| Attached | |
| 2X-Large | |
| Front Zipper with Double Storm Flap | |
| Adjustable webbing, straps with closure; Multi-layer film barrier laminated | |
| 6 to 6.4 ft. | |
| 29 in. | |
| 35.2 in. | |
| 89.4 cm | |
| 28.25 in. | |
| 18 mil |