missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tychem™ CPF 3 Combo Suit
Description
Tychem 5000 fabric is composed of a multi-layer film barrier laminated to a durable 2.0 oz/yd2 polypropylene substrate. Tychem 5000 fabric is strong and durable for rigorous activities and rugged situations involving liquid splash and provides barrier to a broad range of chemicals. Typical Applications: chemical handling, petro-chemical, hazardous materials/waste clean-up, fire departments, industrial hazmat teams, utilities, and domestic preparedness. Commonly used in domestic preparedness for situations where the potential to exposure to chemicals exist.Jacket and bib overall combination ensemble. Components also sold separatelyComes with a jam fit cuff and glove insert for user supplied gloves
Specifications
Specifications
| Certifications/Compliance | ASTM D751, ASTM D1117, ASTM D3787, ASTM D751, ASTM D2582, ASTM D751, 16CFR1610 |
| Color | Tan |
| For Use With (Application) | Chemical handling, petro-chemical, hazardous materials/waste clean-up, fire departments, industrial hazmat teams, utilities, and domestic preparedness |
| Hood Type | Attached |
| Material | Multi-layer Barrier Laminate Film |
| Size | Large |
| Ankle Style | Open |
| Closure Type | Front Zipper with Double Storm Flap |
| Cuff Style | Elastic |
| Description | Adjustable webbing, straps with closure; Multi-layer film barrier laminated |
| Show More |