missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tychem™ 5000 Encapsulated Level B Suit
Description
- Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion
- Manufactured under specifications that do not contain natural rubber latex
- Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.
- Also provides coverage from overhead liquid splash (does not provide vapor protection)
- Taped seams provide strong chemical resistance against heavy liquid splashes
- Sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
- Standard 20 mil PVC face shield
- Rear entry design keeps closure away from direct contact with frontal chemical exposure and provides ability to change air bottle on SCBA without removing the entire garment
- Expanded back to accommodate self-contained air breathing apparatus (SCBA)
- Storm flap covers zipper which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper
- Elastic opening for tighter fit at wrist
- Integrated socks composed of garment material
- D13469623
Specifications
Specifications
| Color | Tan |
| For Use With (Application) | Chemical Handling, Hazmat, Law Enforcement, Domestic Preparedness |
| Material | Tychem |
| Size | 4X-Large |
| Ankle Style | Boot Overflaps |
| Closure Type | Zipper |
| Product Type | Encapsulated Level B Suit |
| Seam Type | Taped |