missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tychem™ 5000 Bib Overall

Catalog No. p-26664
Change view
Click to view available options
Size:
2X-Large
3X-Large
Size (Metric) Chest:
109.85 to 118.74 cm
120 to 128.9 cm
Length (Metric):
182.88 to 195.07 cm
188.97 to 195.07 cm
Length (English) Inseam:
30.75 in.
31.5 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Size (Metric) Chest Length (Metric) Length (English) Inseam
189992596 3X-Large 120 to 128.9 cm 188.97 to 195.07 cm 31.5 in.
296361539 2X-Large 109.85 to 118.74 cm 182.88 to 195.07 cm 30.75 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Should not be worn around open flame, heat source, sparks or ignition sources.
Catalog No. 189992596 Supplier DuPont™ Supplier No. C3360TTN3X000600
Only null left
Should not be worn around open flame, heat source, sparks or ignition sources.

Offers protection from numerous chemicals and other hazardous materials

  • Bib style overall with adjustable webbing straps
  • Typically worn in combination with C3670T TN jacket or in appplications were potential exposure is below waist
  • Manufactured under specifications that do not contain natural rubber latex
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks
  • D13469468,D13469473

Specifications

Closure Type Snap
Color Tan
Material Tyvek
Seam Type Taped
Size 3X-Large
Ankle Style Open
Description Open ankle, adjustable webbing straps with buckle closure; Tychem™ CPF3 Fabric is composed of a multilayer film barrier laminated to a durable 2 oz. Polypropylene substrate
Cuff Style Elastic
For Use With (Application) C3670T TN jacket or in appplications where potential exposure is below waist. Chemical handling, petrochemical, hazardous materials, waste cleanup, fire departments, industrial hazmat teams, utilities and domestic preparedness
Length (English) 6.2 to 6.4 ft.
Length (English) Inseam 31.5 in.
Length (Metric) 188.97 to 195.07 cm
Size (Metric) Chest 120 to 128.9 cm
Thickness 0.45 mm
Weight (English) 0.27 lb.
Weight (Metric) 0.12 kg
Certifications/Compliance ASTM D1117
Disposable Single-use
Show More Show Less