missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tychem™ 5000 Bib Overall

Catalog No. 296361539
Change view
Click to view available options
Size:
2X-Large
3X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
296361539 2X-Large
189992596 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Should not be worn around open flame, heat source, sparks or ignition sources.
Catalog No. 296361539 Supplier DuPont™ Supplier No. C3360TTN2X000600

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Should not be worn around open flame, heat source, sparks or ignition sources.

Offers protection from numerous chemicals and other hazardous materials

  • Bib style overall with adjustable webbing straps
  • Typically worn in combination with C3670T TN jacket or in applications were potential exposure is below waist
  • Manufactured under specifications that do not contain natural rubber latex
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks
  • D13469468, D13469473
  • Open ankle, adjustable webbing straps with buckle closure; Tychem™ CPF3 Fabric is composed of a multilayer film barrier laminated to a durable 2 oz. Polypropylene substrate

Specifications

Closure Type Snap
Color Tan
Material Tyvek
Seam Type Taped
Size 2X-Large
Suspender Adjustable Webbing Straps with Buckle Closure
Ankle Style Open
Cuff Style Elastic
For Use With (Application) Below the Waist Exposure, Chemical Handling, Petrochemical, Hazardous Materials, Waste Cleanup, Fire Departments, Industrial Hazmat Teams, Utilities and Domestic Preparedness
Length (English) 6 to 6.4 ft.
Length (English) Inseam 30.75 in.
Length (Metric) 182.88 to 195.07 cm
Product Type Protective Bib Overall
Size (English) Chest 43.25 to 46.75 in.
Size (Metric) Chest 109.85 to 118.74 cm
Thickness 0.45 mm
Weight (English) 0.27 lb.
Weight (Metric) 0.12 kg
Certifications/Compliance ASTM D1117
Disposable Single-use
Show More Show Less