Learn More
DuPont™ ProShield™ 10 Frock
SMS frocks are designed to be used in a range of industries for nonhazardous dry particle and light splash applications.
Supplier:  DuPont™ PB267SWHXL003000
 View more versions of this product
Catalog No. 19-162-637
Only null left
Explore available promotions
Description
Description
  • Frock provides body-length jacket design with front snap closures (6+1 adjustable) for easy donning and doffing
  • Mandarin collar
  • Extends to knee
  • Set sleeve
  • Knit cuff
  • Enclosed elastic wrist
  • Serged seams
  • Manufactured under specifications that do not contain natural rubber latex
Specifications
Specifications
General Purpose Frock
ProShield
Single-use
Elastic
General Maintenance, Janitorial, Cleaning
Yes
X-Large
White
Snaps
Serged
Mandarin
Product Suggestions
Product Suggestions