missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ ProShield™ 10 Coverall (all)

Catalog No. 19162513
Change view
Click to view available options
Color:
Blue
White
Size:
Medium
Large
X-Large
5X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19162513 Blue 5X-Large
19162501 White Large
19162506 White 5X-Large
19162507 Blue Medium
19162509 Blue X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19162513 Supplier DuPont™ Supplier No. PB120SBU5X002500

Spunbond meltblown spunbond (SMS) coveralls protect against non-hazardous particles and light liquid splashes in a variety of industries.

  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Laydown collar
  • Open wrists
  • Open ankles
  • Manufactured under specifications that do not contain natural rubber latex

Specifications

Product Type General Purpose Coveralls
Size 5X-Large
Gender Unisex
Material ProShield
Color Blue
Disposable Single-use
Waist Style Elastic
Ankle Style Open
Cuff Style Open
Neck Style Collared
Hood Type Unhooded
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Collar Type Laydown
Model PB120S
For Use With (Application) General Maintenance, Janitorial, Cleaning
Quantity 25/Cs.
Show More Show Less