Learn More
DuPont™ ProShield™ 10 Coverall (all)
Spunbond meltblown spunbond (SMS) coveralls protect against non-hazardous particles and light liquid splashes in a variety of industries.
Supplier:  DuPont™ PB120SWH5X002500
 View more versions of this product
Catalog No. 19162506
Only null left
Explore available promotions
Description
Description
Spunbond meltblown spunbond (SMS) coveralls protect against non-hazardous particles and light liquid splashes in a variety of industries.
  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Laydown collar
  • Open wrists
  • Open ankles
  • Manufactured under specifications that do not contain natural rubber latex
Specifications
Specifications
General Purpose Coveralls
Unisex
White
Elastic
Open
Unhooded
Serged
Laydown
General Maintenance, Janitorial, Cleaning
5X-Large
ProShield
Single-use
Open
Collared
Front Zipper
Non-sterile
PB120S
25/Cs.
Product Suggestions
Product Suggestions