Learn More
DuPont™ ProShield™ 10 Coverall (all)
Spunbond meltblown spunbond (SMS) coveralls protect against non-hazardous particles and light liquid splashes in a variety of industries.
Supplier:  DuPont™ PB125SBUXL002500
 View more versions of this product
Catalog No. 19-162-537
Only null left
Explore available promotions
Description
Description
Spunbond meltblown spunbond (SMS) coveralls protect against non-hazardous particles and light liquid splashes in a variety of industries.
  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Laydown collar
  • Elastic opening for tighter fit at wrist
  • Elastic opening for tighter fit at ankle
  • Manufactured under specifications that do not contain natural rubber latex
Specifications
Specifications
General Purpose Coveralls
Unisex
Blue
Elastic
Elastic
Unhooded
Serged
Laydown
General Maintenance, Janitorial, Cleaning
X-Large
ProShield
Single-use
Elastic
Collared
Front Zipper
Non-sterile
PB125S
25/Cs.
Product Suggestions
Product Suggestions